Anti-IgG Goat Polyclonal Antibody (CF™ 555)

Supplier: Biotium

20230 20230-1
89138-780EA 260.49 USD
89138-780 89138-782
Anti-IgG Goat Polyclonal Antibody (CF™ 555)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 555
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR