Anti-IgG Bovine Polyclonal Antibody (CF™ 488A)

Supplier: Biotium

20293 20293-1
71003-912EA 260.49 USD
71003-912 71003-914
Anti-IgG Bovine Polyclonal Antibody (CF™ 488A)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 488A
Epitope: Heavy and Light chain
Host: Bovine
Isotype: IgG
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR