Anti-IgG1 Goat Polyclonal Antibody (CF™ 488A)

Supplier: Biotium

20246 20246-1
71004-022EA 279.11 USD
71004-022 71004-024
Anti-IgG1 Goat Polyclonal Antibody (CF™ 488A)
Antibodies
Type: Secondary
Antigen: IgG1
Clonality: Polyclonal
Clone:
Conjugation: CF™ 488A
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR