Anti-IgG Goat Polyclonal Antibody (CF™ 488A)

Supplier: Biotium

20017 20017-1
89138-402EA 217.08 USD
89138-402 71004-194
Anti-IgG Goat Polyclonal Antibody (CF™ 488A)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 488A
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR