Anti-IgG Goat Polyclonal Antibody (CF™ 488A)

Supplier: Biotium

20011 20011-1
89138-384EA 193.15 USD
89138-384 10018-236
Anti-IgG Goat Polyclonal Antibody (CF™ 488A)
Antibodies
This conjugate is prepared by labeling F(ab’)2 fragment of goat anti-mouse IgG (H+L) with CF™488A dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Green fluorescent CF™488A (Ex/Em 490/515 nm) is a superior alternative to FITC (or fluorescein) owing to its exceptional brightness and photostability.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 488A
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR