Anti-IgG Goat Polyclonal Antibody (CF™ 488A)

Supplier: Biotium

20010 20010-1
89138-382EA 268.81 USD
89138-382 71004-190
Anti-IgG Goat Polyclonal Antibody (CF™ 488A)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 488A
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR