Anti-IgG Goat Polyclonal Antibody (CF™ 405M)

Supplier: Biotium

20182 20182-1
89138-756EA 260.49 USD
89138-756 89138-758
Anti-IgG Goat Polyclonal Antibody (CF™ 405M)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 405M
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR