Anti-IgG Donkey Polyclonal Antibody (CF™ 405S)

Supplier: Biotium

20356-500UL 20356-50UL
89411-132EA 260.49 USD
89411-132 89411-546
Anti-IgG Donkey Polyclonal Antibody (CF™ 405S)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 405S
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR