Anti-IgG Goat Polyclonal Antibody (CF™ 350)

Supplier: Biotium

20145 20145-1
89410-882EA 172.63 USD
89410-882 10018-254
Anti-IgG Goat Polyclonal Antibody (CF™ 350)
Antibodies
This conjugate is prepared by labeling F(ab’)2 fragment of goat anti-mouse IgG (H+L) with CF™350 dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Blue fluorescent CF™350 (Ex/Em 347/448 nm) is more water-soluble, more photostable and at least 50% more fluorescent on proteins compared to the traditional blue dye AMCA.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 350
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR