Anti-IgG Donkey Polyclonal Antibody (CF™ 350)

Supplier: Biotium

20148 20148-1
89138-714EA 260.49 USD
89138-714 89138-716
Anti-IgG Donkey Polyclonal Antibody (CF™ 350)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 350
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR