Anti-CD107a (LAMP-1) Rat Monoclonal Antibody [clone: 1D4B]

Supplier: Biolegend
121601 121602
121601-BLEA 93.4 USD
121601-BL 121602-BL
Anti-CD107a (LAMP-1) Rat Monoclonal Antibody [clone: 1D4B]
Antibodies
Purified anti-mouse CD107a (LAMP-1) [1D4B]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IF, IHC, IP; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD107a (LAMP-1)
Clonality: Monoclonal
Clone: 1D4B
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR