Anti-TNF-a Mouse Monoclonal Antibody [clone: MAb1]

Supplier: Biolegend
502801 502802
502801-BLEA 93.4 USD
502801-BL 502802-BL
Anti-TNF-a Mouse Monoclonal Antibody [clone: MAb1]
Antibodies
Purified anti-human TNF-α [MAb1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ELISA Capture, Neut, WB; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA capture applications, a concentration range of 0.25-1 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of TNF-α recombinant protein ranging from 500 to 4 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TNF-a
Clonality: Monoclonal
Clone: MAb1
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR