Anti-IL-6 Rat Monoclonal Antibody [clone: MQ2-13A5]

Supplier: Biolegend
501101 501102
501101-BLEA 97.66 USD
501101-BL 501102-BL
Anti-IL-6 Rat Monoclonal Antibody [clone: MQ2-13A5]
Antibodies
Purified anti-human IL-6 [MQ2-13A5]; Isotype: Rat IgG1, κ; Reactivity: Human; Apps: ELISA Capture, CyTOF®; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA capture applications, a concentration range of 0.5-2.0 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of IL-6 recombinant protein ranging from 1000 to 8 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-6
Clonality: Monoclonal
Clone: MQ2-13A5
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR