Anti-TCR-b chain Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: H57-597]

Supplier: Biolegend
109221 109222
109221-BLEA 109.1 USD
109221-BL 109222-BL
Anti-TCR-b chain Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: H57-597]
Antibodies
PE/Cy7 anti-mouse TCR β chain [H57-597]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TCR-b chain
Clonality: Monoclonal
Clone: H57-597
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR