Anti-CD45 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: 30-F11]

Supplier: Biolegend
103109 103110
103109-BLEA 91.98 USD
103109-BL 103110-BL
Anti-CD45 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: 30-F11]
Antibodies
PE/Cy5 anti-mouse CD45 [30-F11]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: 30-F11
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR