Anti-CD134 (OX-40) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: OX-86]

Supplier: Biolegend
119409 119410
119409-BLEA 142.5 USD
119409-BL 119410-BL
Anti-CD134 (OX-40) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: OX-86]
Antibodies
PE anti-mouse CD134 (OX-40) [OX-86]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD134 (OX-40)
Clonality: Monoclonal
Clone: OX-86
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR