Anti-Ly-6G/Ly-6C (Gr-1) Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: RB6-8C5]

Supplier: Biolegend
108429 108430
108429-BLEA 110.7 USD
108429-BL 108430-BL
Anti-Ly-6G/Ly-6C (Gr-1) Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: RB6-8C5]
Antibodies
Pacific Blue™ anti-mouse Ly-6G/Ly-6C (Gr-1) [RB6-8C5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Ly-6G/Ly-6C (Gr-1)
Clonality: Monoclonal
Clone: RB6-8C5
Conjugation: PB (Pacific Blue)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR