Anti-IgG2b, k Isotype Ctrl Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RTK4530]

Supplier: Biolegend
400633 400605 400606 400634
400633-BLEA 47.73 USD
400633-BL 400605-BL 400606-BL 400634-BL
Anti-IgG2b, k Isotype Ctrl Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RTK4530]
Antibodies
FITC Rat IgG2b, κ Isotype Ctrl [RTK4530]; Isotype: Rat IgG2b, κ; Reactivity: Rat; Apps: FC, ICFC; Size: 100 tests

Each lot of this rat IgG2b, κ isotype control antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, use the isotype control at the same concentration as your primary antibody. Use our Concentration Lookup tool to find the exact concentrations of your lots of product.

Type: Control
Antigen: IgG2b, k Isotype Ctrl
Clonality: Monoclonal
Clone: RTK4530
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR