Anti-CD44 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: IM7]

Supplier: Biolegend
103005 103006 103021 103022
103005-BLEA 61.3 USD
103005-BL 103006-BL 103021-BL 103022-BL
Anti-CD44 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: IM7]
Antibodies
FITC anti-mouse/human CD44 [IM7]; Isotype: Rat IgG2b, κ; Reactivity: Mouse, Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus, Squirrel Monkey, Horse (Equine), Cattle (Bovine, Cow), Swine (Pig, Porcine), Dog (Canine), Cat (Feline); Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining using the μg size, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: IM7
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse;Human;Cross-Reactivity: Chimpanzee;Baboon;Cynomolgus;Rhesus;Squirrel Monkey;Horse (Equine);Cattle (Bovine;Cow);Swine (Pig;Porcine);Dog (Canine);Cat (Feline)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR