Anti-IgG2a, k Isotype Ctrl Rat Monoclonal Antibody (Biotin) [clone: RTK2758]

Supplier: Biolegend
400503 400504
400503-BLEA 83.56 USD
400503-BL 400504-BL
Anti-IgG2a, k Isotype Ctrl Rat Monoclonal Antibody (Biotin) [clone: RTK2758]
Antibodies
Biotin Rat IgG2a, κ Isotype Ctrl [RTK2758]; Isotype: Rat IgG2a, κ; Reactivity: KLH; Apps: FC, ICFC; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest.

Type: Control
Antigen: IgG2a, k Isotype Ctrl
Clonality: Monoclonal
Clone: RTK2758
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: KLH
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR