Anti-IgG2a Rat Monoclonal Antibody (Biotin) [clone: RMG2a-62]

Supplier: Biolegend
407103 407104
407103-BLEA 93.89 USD
407103-BL 407104-BL
Anti-IgG2a Rat Monoclonal Antibody (Biotin) [clone: RMG2a-62]
Antibodies
Biotin anti-mouse IgG2a [RMG2a-62]; Isotype: Rat IgG; Reactivity: Mouse; Apps: ELISA, FC; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA. For ELISA detection, the suggested use of this reagent is 1.0 μg per ml. It's recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IgG2a
Clonality: Monoclonal
Clone: RMG2a-62
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR