Anti-CD11b Rat Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: M1/70]

Supplier: Biolegend
101225 101226
101225-BLEA 131.3 USD
101225-BL 101226-BL
Anti-CD11b Rat Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: M1/70]
Antibodies
APC/Cy7 anti-mouse/human CD11b [M1/70]; Isotype: Rat IgG2b, κ; Reactivity: Mouse, Human, Cross-Reactivity: Cynomolgus, Rhesus, Baboon, Chimpanzee,Rabbit (Lapine); Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD11b
Clonality: Monoclonal
Clone: M1/70
Conjugation: APC (Allophycocyanin)/Cy7®
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse;Human;Cross-Reactivity: Cynomolgus;Rhesus;Baboon;Chimpanzee,Rabbit (Lapine)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR