Anti-Mac-3 Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: M3/84]

Supplier: Biolegend
108511 108512
108511-BLEA 113.7 USD
108511-BL 108512-BL
Anti-Mac-3 Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: M3/84]
Antibodies
Alexa Fluor® 647 anti-mouse CD107b (Mac-3) [M3/84]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC, IF, IHC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. For immunofluorescence microscopy, a concentration range of 5-10 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Mac-3
Clonality: Monoclonal
Clone: M3/84
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR