Anti-CD90 (Thy1) Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: 5E10]

Supplier: Biolegend
328115 328116
328115-BLEA 139.6 USD
328115-BL 328116-BL
Anti-CD90 (Thy1) Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: 5E10]
Antibodies
Alexa Fluor® 647 anti-human CD90 (Thy1) [5E10]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Pigtailed Macaque, Rhesus, Swine (Pig, Porcine); Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD90 (Thy1)
Clonality: Monoclonal
Clone: 5E10
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Pigtailed Macaque;Rhesus;Swine (Pig;Porcine)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR