Anti-MAdCAM-1 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: MECA-367]

Supplier: Biolegend
120707 120708
120707-BLEA 113.7 USD
120707-BL 120708-BL
Anti-MAdCAM-1 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: MECA-367]
Antibodies
Alexa Fluor® 488 anti-mouse MAdCAM-1 [MECA-367]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: MAdCAM-1
Clonality: Monoclonal
Clone: MECA-367
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR