Anti-CD10 Calla Mouse Monoclonal Antibody [clone: 56C6]

Supplier: Diagnostic Biosystems

MOB240 MOB240-05
99990-468EA 1022.54 USD
99990-468 95024-310
Anti-CD10 Calla Mouse Monoclonal Antibody [clone: 56C6]
Antibodies
Type:
Antigen: CD10 Calla
Clonality: Monoclonal
Clone: 56C6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR