OptiTDS™, Pancreas Tissue Dissociation System, CHI Scientific
Supplier: CHI Scientific
4-20371 4-20372 4-20373 4-20374 4-20375 4-20376 4-20377 4-20378 4-20379 4-20380
10783-250EA 340.58 USD
10783-250 10786-232 10786-234 10786-236 10786-238 10786-240 10786-242 10786-244 10786-246 10786-248
OptiTDS™, Pancreas Tissue Dissociation System, CHI Scientific
Protein Extraction Protein Extraction Kits
OptiTDS™ is a proprietary Tissue Dissociation system which enzymatically digests extracellular connective fibers without harming the cells themselves
Stability/Storage: The reagents contained in this system are stable at room temperatures for normal shipping periods, but the kit should be used immediately or the package should be stored at -20oC upon arrival. The reagents contained in this kit are stable and effective for up to 6 months at -20oC.
Each OptiTDS™ system contains concentrated tissue dissociation enzymes and reagents which are added to an included buffer system to produce a working solution that is capable of treating 3-5 g of tissue. Each tissue dissociation system is customized to be tissue-specific and cell-specific to obtain the best results for each type of primary cell.