Baume/SG Dual Scale Hydrometers, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
52122-0026 52122-2443 52122-4255 52122-5465 52122-0072
10169-750EA 47.03 USD
10169-750 10169-752 10169-754 10169-756 10169-758
Baume/SG Dual Scale Hydrometers, Kimble Chase
Hydrometers
Serial number is printed on the scale and a certificate of traceability is included
The hydrometer meets or exceeds the requirements in ASTM specification E100, and it is calibrated to ASTM specification E126 to ensure accuracy. The certificate of traceability indicates that the hydrometers are manufactured using devices calibrated using NIST traceable standards. The ballast is made from steel pellets and a binder, and it is free of heavier metals.