5-Bromo-2-methoxypyridine-3-carboxaldehyde ≥99%

Supplier: BeanTown Chemical

Synonyms: 5-Bromo-2-methoxynicotinaldehyde

225990-1G 225990-5G
BT225990-1GEA 206 USD
BT225990-1G BT225990-5G
5-Bromo-2-methoxypyridine-3-carboxaldehyde ≥99%
5-Bromo-2-methoxypyridine-3-carboxaldehyde

CAS: 103058-87-3; MDL No: MFCD05663521 Solid; Molecular Formula: C7H6BrNO2; MW: 216.03 Melting Point: 93-100° Air Sensitive


  • Air Sensitive
  • Solid

Formula: C₇H₆BrNO₂
MW: 216.034 g/mol
Melting Pt: 93…100 °C
MDL Number: MFCD05663521
CAS Number: 103058-87-3

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR