7-Bromo-1-tetralone ≥97%

Supplier: BeanTown Chemical

Synonyms: 7-Bromo-3,4-dihydronaphthalen-1(2H)-one

225980-1G 225980-5G
BT225980-1GEA 167 USD
BT225980-1G BT225980-5G
7-Bromo-1-tetralone ≥97%
7-Bromo-1-tetralone

CAS: 32281-97-3; MDL No: MFCD02179287 Solid; Molecular Formula: C10H9BrO; MW: 225.08 Melting Point: 75-80°


  • Solid

Formula: C₁₀H₉BrO
Melting Pt: 75…80 °C
MDL Number: MFCD02179287
CAS Number: 32281-97-3

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR