Ammonium sulphamate 98,0 ACS
Supplier: BeanTown Chemical
Synonyms:
Ammonium sulphamidate, Ammonium amidosulphonate
225965-2KG
225965-500G
BT225965-2KGEA
247
USD
BT225965-2KG
BT225965-500G
Ammonium sulphamate 98,0 ACS
Ammonium sulphamate
CAS: 7773-06-0; EC No: 231-871-7; MDL No: MFCD00011429; RTECS: WO6125000
Crystalline; Linear Formula: NH4SO3NH2; Molecular Formula: H6N2O3S; MW: 114.12
Melting Point: 131-135°
Air Sensitive, Moisture Sensitive
- RTECS: WO6125000
- Air Sensitive, Moisture Sensitive
- Crystalline
Formula:
H₆N₂O₃S MW: 114.13 g/mol Melting Pt: 131.1 |
MDL Number:
MFCD00011429 CAS Number: 7773-06-0 EINECS: 231-871-7 Merck Index: 12,00589 |
Specification Test Results
Assay | 98.0% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...