O-Methylisourea hemisulfate ≥99%

Supplier: BeanTown Chemical

Synonyms: O-Methylisouronium sulphate

225930-1KG 225930-250G 225930-50G
BT225930-1KGEA 577 USD
BT225930-1KG BT225930-250G BT225930-50G
O-Methylisourea hemisulfate ≥99%
O-Methylisourea hemisulfate

CAS: 52328-05-9; EC No: 257-851-8; MDL No: MFCD00040594 Crystalline; Linear Formula: H2NC(OCH3)=NH·0.5H2SO4; Molecular Formula: C2H6N2O·0.5H2SO4; MW: 123.12 Melting Point: 163-167°


  • Crystalline

Formula: C₄H₁₄N₄O₆S
MW: 246.24 g/mol
Melting Pt: 163…167 °C
MDL Number: MFCD00040594
CAS Number: 52328-05-9
EINECS: 257-851-8

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR