3-Amino-5-nitrobenzoic acid ≥97%

Supplier: BeanTown Chemical

225890-1G 225890-25G 225890-5G
BT225890-1GEA 37 USD
BT225890-1G BT225890-25G BT225890-5G
3-Amino-5-nitrobenzoic acid ≥97%
3-Amino-5-nitrobenzoic acid

CAS: 618-84-8; MDL No: MFCD00017025 Powder; Molecular Formula: C7H6N2O4 ; MW: 182.14


  • Powder

Formula: C₇H₆N₂O₄
MDL Number: MFCD00017025
CAS Number: 618-84-8

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR