Cyanuric acid ≥99%
Supplier: BeanTown Chemical
Synonyms:
1,3,5-Triazinetrion, 1,3,5-Triazine-2,4,6-triol, 1,3,5-Triazinane-2,4,6-trione, Isocyanuric acid
225870-1KG
225870-250G
BT225870-1KGEA
102
USD
BT225870-1KG
BT225870-250G
Cyanuric acid ≥99%
Cyanuric acid
CAS: 108-80-5; EC No: 203-618-0; MDL No: MFCD00082990; RTECS: XZ1800000
Powder; Molecular Formula: C3H3N3O3; MW: 129.08
Melting Point:
- RTECS: XZ1800000
- Powder
Formula:
C₃H₃N₃O₃ MW: 129.08 g/mol Melting Pt: >360 °C |
MDL Number:
MFCD00082990 CAS Number: 108-80-5 EINECS: 203-618-0 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...