4-Bromobiphenyl (PBB No. 3) ≥98%
Supplier: BeanTown Chemical
Synonyms:
4-Bromodiphenyl, PBB No. 3, 4-Bromobiphenyl
225860-10G
225860-250G
225860-50G
BT225860-10GEA
30
USD
BT225860-10G
BT225860-250G
BT225860-50G
4-Bromobiphenyl (PBB No. 3) ≥98%
4-Bromobiphenyl (PBB No. 3)
CAS: 92-66-0; EC No: 202-176-6; MDL No: MFCD00000100; RTECS: DV1750100
Crystalline; Molecular Formula: C12H9Br; MW: 233.10
Melting Point: 82-86°; Boiling Point: 310°; Flash point: 100°C (212°F)
- RTECS: DV1750100
- Crystalline
Formula:
C₁₂H₉Br MW: 233.11 g/mol Boiling Pt: 310 °C Melting Pt: 82…86 °C Flash Pt: 100 °C (212 °F) |
MDL Number:
MFCD00000100 CAS Number: 92-66-0 EINECS: 202-176-6 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...