(Triphenylphosphoranylidene)acetaldehyde, (max. 3% H₂O) ≥97%

Supplier: BeanTown Chemical

225845-100G 225845-10G 225845-25G
BT225845-100GEA 479 USD
BT225845-100G BT225845-10G BT225845-25G
(Triphenylphosphoranylidene)acetaldehyde, (max. 3% H₂O) ≥97%
(Triphenylphosphoranylidene)acetaldehyde

CAS: 2136-75-6; EC No: 218-375-6; MDL No: MFCD00006994 Crystalline; Linear Formula: (C6H5)3P=CHCHO; Molecular Formula: C20H17OP; MW: 304.32 Melting Point: 185-188° Air Sensitive


  • Air Sensitive
  • Crystalline

Formula: C₂₀H₁₇OP
Melting Pt: 185…188 °C
MDL Number: MFCD00006994
CAS Number: 2136-75-6
EINECS: 218-375-6

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR