1-Naphthyl boronic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: 1-Naphthaleneboronic acid, Naphthalene-1-boronic acid, Naphthalen-1-ylboronic acid

225755-1G 225755-25G 225755-5G
BT225755-1GEA 38 USD
BT225755-1G BT225755-25G BT225755-5G
1-Naphthyl boronic acid ≥97%
1-Naphthyl boronic acid

CAS: 13922-41-3; MDL No: MFCD00019722 Powder; Molecular Formula: C10H9BO2 ; MW: 171.99 Melting Point: 208-214°


  • Powder

Formula: C₁₀H₉BO₂
MW: 171.99 g/mol
Melting Pt: 208…214 °C
MDL Number: MFCD00019722
CAS Number: 13922-41-3

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR