Poly(2-ethyl-2-oxazoline), M.W. ~200,000

Supplier: BeanTown Chemical

225725-100G 225725-2.5KG 225725-500G
BT225725-100GEA 30 USD
BT225725-100G BT225725-2.5KG BT225725-500G
Poly(2-ethyl-2-oxazoline), M.W. ~200,000
Poly(2-ethyl-2-oxazoline), M.W. ~200,000

CAS: 25805-17-8; MDL No: MFCD00134377 Solid; Linear Formula: [-N(COC2H5)CH2CH2-]n ; MW ≈200,000 Density (g/mL): 1.14 Moisture Sensitive


  • Moisture Sensitive
  • Solid

Formula: (C₅H₉NO)x
MW: 200000 g/mol
Density: 1.14 g/mL
CAS Number: 25805-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR