Ammonium metatungstate hydrate
Supplier: BeanTown Chemical
Synonyms:
Ammonium meta-tungsten oxide hydrate
225635-100G
225635-25G
225635-500G
BT225635-100GEA
77
USD
BT225635-100G
BT225635-25G
BT225635-500G
Ammonium metatungstate hydrate
Ammonium metatungstate hydrate
CAS: 12028-48-7
EC No: 234-733-4
MDL No: MFCD00042673
Powder
Melting Point: 100° (decomposes)
- Powder
Formula:
(NH₄)₆W₁₂O₃₉·xH₂O MW: 2938.41 g/mol (anhy) Melting Pt: 100 °C (dec.) |
MDL Number:
MFCD00042673 CAS Number: 12028-48-7 EINECS: 234-733-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...