Polyaniline, emeraldine base

Supplier: BeanTown Chemical

Synonyms: 5-Bromo-4-methyl-1H-1,2,3-triazole hydrochloride

225615-1G 225615-5G
BT225615-1GEA 30 USD
BT225615-1G BT225615-5G
Polyaniline, emeraldine base
Polyaniline, emeraldine base

CAS: 25233-30-1; MDL No: MFCD00241446 Powder; Linear Formula: [-C6H5NH-]n; MW ≈15,000 Melting Point: 200° (decomposes)


  • MW ≈15,000

Formula: C₃H₅BrClN₃
Melting Pt: >200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00241446
CAS Number: 25233-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR