Diyttrium trioxide ≥99.995% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Yttrium oxide, Yttrium (III) oxide
225600-100G
225600-25G
225600-500G
BT225600-100GEA
488
USD
BT225600-100G
BT225600-25G
BT225600-500G
Diyttrium trioxide ≥99.995% (trace metals basis)
Diyttrium trioxide
CAS: 1314-36-9; EC No: 215-233-5; MDL No: MFCD00011473
25-50nm APS Powder; Linear Formula: Y2O3; MW: 225.81
Melting Point: 2410°; Boiling Point: 4300°
Density (g/mL): 5.01
Air Sensitive
- Air Sensitive
- 25-50nm APS Powder
Formula:
Y₂O₃ MW: 225.81 g/mol Boiling Pt: 4300 °C Melting Pt: 2410 °C Density: 5.01 g/mL |
MDL Number:
MFCD00011473 CAS Number: 1314-36-9 EINECS: 215-233-5 |
Specification Test Results
Assay | 99.995% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...