D-(+)-Trehalose dihydrate ≥99%

Supplier: BeanTown Chemical

Synonyms: Mycose, α,α-Trehalose, 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside, α-D-Glucopyranosyl-α-D-glucopyranoside, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol) dihydrate

225575-100G 225575-25G 225575-5G
BT225575-100GEA 311.38 USD
BT225575-100G BT225575-25G BT225575-5G
D-(+)-Trehalose dihydrate ≥99%
D-(+)-Trehalose dihydrate

CAS: 6138-23-4; EC No: 202-739-6; MDL No: MFCD00071594 Solid; Molecular Formula: C12H22O11·2H2O; MW: 378.33 Melting Point: 97-99° Hygroscopic


  • Hygroscopic
  • Solid

Formula: C₁₂H₂₂O₁₁·2H₂O
MW: 378.33 g/mol
Melting Pt: 97…99 °C
MDL Number: MFCD00071594
CAS Number: 6138-23-4
EINECS: 202-739-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af741ee934bfe7c82395d31c21e2c19f0738d0f0&vid=59183c854dcfa367e15d46861ef4b9a7028540a9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR