(S)-1-(tert-Butoxycarbonyl)-4,4-difluoro-2-pyrrolidinecarboxylic acid ≥97%
Supplier: BeanTown Chemical
Synonyms:
(2S)-1-[(tert-butoxy)carbonyl]-4,4-difluoropyrrolidine-2-carboxylic acid, (S)-1-(tert-Butoxycarbonyl)-4,4-difluoropyrrolidine-2-carboxylic acid, N-Boc-4,4-difluoro-L-proline, (S)-1-Boc-4,4-difluoro-2-pyrrolidinecarboxylic Acid, N-(tert-Butoxycarbonyl)-4,4-difluoro-L-proline
225560-1G
225560-250MG
BT225560-1GEA
321
USD
BT225560-1G
BT225560-250MG
(S)-1-(tert-Butoxycarbonyl)-4,4-difluoro-2-pyrrolidinecarboxylic acid ≥97%
(S)-1-(tert-Butoxycarbonyl)-4,4-difluoro-2-pyrrolidinecarboxylic acid
CAS: 203866-15-3; MDL No: MFCD03094917
Solid; Molecular Formula: C10H15F2NO4; MW: 251.24
Melting Point: 120-124°
Optical Rotation: [α]22/D -69.0° in chloroform
- Optical rotation: [α]22/D -69.0° in chloroform
- Solid
Formula:
C₁₀H₁₅F₂NO₄ MW: 251.23 g/mol Melting Pt: 120…124 °C |
MDL Number:
MFCD03094917 CAS Number: 203866-15-3 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...