2-Methyl-1-phenyl-1-propanol ≥98%

Supplier: BeanTown Chemical

Synonyms: α-Isopropylbenzyl alcohol, 2-Methyl-1-phenylpropan-1-ol

225545-25G 225545-5G
BT225545-25GEA 374 USD
BT225545-25G BT225545-5G
2-Methyl-1-phenyl-1-propanol ≥98%
2-Methyl-1-phenyl-1-propanol

CAS: 611-69-8; EC No: 210-274-5; MDL No: MFCD00065000 Liquid; Molecular Formula: C10H14O; MW: 150.22 Flash point: 86°C (187°F) Density (g/mL): 0.964


  • Liquid

Formula: C₆H₅CH(OH)CH(CH₃)₂
MW: 150.22 g/mol
Density: 0.964 g/mL
Flash Pt: 86 °C (187 °F)
MDL Number: MFCD00065000
CAS Number: 611-69-8
EINECS: 210-274-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=954485764810b5712627a8e76df6a8dd392a6e2a&vid=954485764810b5712627a8e76df6a8dd392a6e2a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR