2,3,4-Trihydroxybenzaldehyde ≥98%

Supplier: BeanTown Chemical

225540-25G 225540-5G
BT225540-25GEA 203 USD
BT225540-25G BT225540-5G
2,3,4-Trihydroxybenzaldehyde ≥98%
2,3,4-Trihydroxybenzaldehyde

CAS: 2144-08-3; EC No: 218-404-2; MDL No: MFCD00003325; RTECS: CU8439800 Powder; Molecular Formula: C7H6O4; MW: 154.12 Melting Point: 159-161° Air Sensitive


  • RTECS: CU8439800
  • Air Sensitive
  • Powder

Formula: C₇H₆O₄
MW: 154.12 g/mol
Melting Pt: 159…161 °C
MDL Number: MFCD00003325
CAS Number: 2144-08-3
EINECS: 218-404-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR