Samarium ≥99.9% (trace metals basis excluding Ta), rods Ø 6.35 mm
Supplier: BeanTown Chemical
225515-10MM
225515-25MM
225515-50MM
BT225515-10MMEA
100
USD
BT225515-10MM
BT225515-25MM
BT225515-50MM
Samarium ≥99.9% (trace metals basis excluding Ta), rods Ø 6.35 mm
Samarium
CAS: 7440-19-9; EC No: 231-128-7; MDL No: MFCD00011233
Solid, cast rod, diameter is approximate; Molecular Formula: Sm; MW: 150.36
Melting Point: 1074°; Boiling Point: 1794°
Density (g/mL): 7.47
- Solid, cast rod, diameter is approximate
Formula:
Sm Boiling Pt: 1794 °C Melting Pt: 1074 °C Density: 7.47 g/mL |
MDL Number:
MFCD00011233 CAS Number: 7440-19-9 EINECS: 231-128-7 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...