Sodium diethyldithiocarbamate trihydrate ACS
Supplier: BeanTown Chemical
CAS: 20624-25-3; EC No: 205-710-6; MDL No: MFCD00150617; RTECS: EZ6550000
Crystalline; Linear Formula: (CH2CH2)2NCSSNa·3H2O; Molecular Formula: C5H10NNaS·3H2O; MW: 225.31
Melting Point: 95-99°
Density (g/mL): 1.100
Hygroscopic
- RTECS: EZ6550000
- Hygroscopic
- Crystalline
Formula:
(C₂H₅)₂NCSSNa·3H₂O Melting Pt: 95…99 °C Density: 1.100 g/mL |
MDL Number:
MFCD00150617 CAS Number: 20624-25-3 EINECS: 205-710-6 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...