4-Bromophenylboronic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: 4-Bromobenzeneboronic acid, (4-Bromophenyl)boronic acid, Dihydroxy(4-bromophenyl)borane, (4-Bromophenyl)metaboric acid

225475-1G 225475-25G 225475-5G
BT225475-1GEA 22 USD
BT225475-1G BT225475-25G BT225475-5G
4-Bromophenylboronic acid ≥97%
4-Bromophenylboronic acid

CAS: 5467-74-3; EC No: 226-779-9; MDL No: MFCD00002104; RTECS: CY8650000 Crystalline/Powder; Molecular Formula: C6H6BBrO2; MW: 200.83 Melting Point: 284-288°


  • RTECS: CY8650000
  • Crystalline/Powder

Formula: C₆H₆BBrO₂
MW: 200.83 g/mol
Melting Pt: 284…288 °C
MDL Number: MFCD00002104
CAS Number: 5467-74-3
EINECS: 226-779-9

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR