2-(Dicyclohexylphosphino)-2',4',6'-triisopropyl-1,1'-biphenyl ≥98%
Supplier: BeanTown Chemical
Synonyms:
XPhos, 2-(Dicyclohexylphosphino)-2',4',6'-tri-i-propyl-1,1'-biphenyl, 2-Dicyclohexylphosphino-2',4',6'-triisopropylbiphenyl
225460-1G
225460-25G
225460-5G
BT225460-1GEA
67
USD
BT225460-1G
BT225460-25G
BT225460-5G
2-(Dicyclohexylphosphino)-2',4',6'-triisopropyl-1,1'-biphenyl ≥98%
2-(Dicyclohexylphosphino)-2',4',6'-triisopropyl-1,1'-biphenyl
CAS: 564483-18-7; MDL No: MFCD04117682
Crystalline/Powder; Molecular Formula: C33H49P; MW: 476.72
Melting Point: 187-190°
Air Sensitive
- Air Sensitive
- Crystalline/Powder
Formula:
C₃₃H₄₉P Melting Pt: 187…190 °C |
MDL Number:
MFCD04117682 CAS Number: 564483-18-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...