2-Bromo-m-xylene ≥98%
Supplier: BeanTown Chemical
CAS: 576-22-7; EC No: 209-397-7; MDL No: MFCD00000075; RTECS: CY9020000
Liquid; Molecular Formula: C8H9Br; MW: 185.06
Melting Point: -10°; Boiling Point: 206°; Flash point: 74°C (165°F)
Density (g/mL): 1.389; Refractive Index: 1.555
- RTECS: CY9020000
- Refractive index: 1.555
- Liquid
Formula:
C₈H₉Br Boiling Pt: 206 °C Melting Pt: –10 °C Density: 1.389 g/mL Flash Pt: 74 °C (165 °F) |
MDL Number:
MFCD00000075 CAS Number: 576-22-7 EINECS: 209-397-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...